Learn More
Hirschmann™ Polypropylene Analytical Funnel
Product Code. 13198139
Change view
Click to view available options
Diameter (Metric)Top:
30mm
50mm
Length (Metric) Overall:
45mm
85mm
Unit Size:
Pack of 24
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top Length (Metric) Overall unitSize
13198139 50mm 85mm Pack of 24
13168139 30mm 45mm Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13198139 Supplier Hirschmann™ Supplier No. 7750104
Only null left
Add to basket
Add to basket
Transfer chemical samples. Hirschmann™ Polypropylene Analytical Funnels are perfect for use in Analytical Chemistry application. Funnels feature a long narrow stem which makes them ideal for use with liquids.
  • Polypropylene
  • Transparent
Specifications
Diameter (Metric)Top 50mm
Diameter (Metric) Stem 7mm
Material Polypropylene
Length (Metric) Stem 43mm
Length (Metric) Overall 85mm