Learn More
Fisherbrand™ Pasteur-plast Pipets
Product Code. 11577722 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Volume (Metric):
1 mL
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11577722 1 mL Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11577722
Estimated Shipment: 01-12-2026
Use these pipettes for simple and safe liquid handling. Fisherbrand™ Pasteur-plast Pipets are inert to biological fluids and most acids. The low affinity of the surface reduces the loss of cells and valuable proteins through binding.
Cross-contamination and carry-over simply cannot occur so that tedious clean-ups are unnecessary. In addition, there is no danger of glass breakage with the resulting danger of infectious material escaping.
  • New soft, elastic bulb
  • Unbreakable – thus reduced risk of glass injury
  • Burr-free orifice for neat, clean and precise dispensing
  • Embossed, external graduations
  • BSE-free can be used for food
Specifications
Material Polypropylene
Sterility Non-sterile
For Use With (Application) Food analysis
Includes Capillary aspiration tip
Quantity 500 Pack
Volume (Metric) 1 mL
Length (Metric) 150 mm
Type Pasteur Pipet