missing translation for 'onlineSavingsMsg'
Learn More

Polyphosphoric Acid, Sigma-Aldrich™

Product Code. 15899470
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15899470 100 mL 100mL
15809480 500 mL 500mL
15819480 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15899470 Supplier Sigma Aldrich Supplier No. 8074710100
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

Polyphosphoric acid (PPA) is a strong mineral acid that combines orthophosphoric acid and linear phosphoric acids. It is widely used in organic synthesis because of its powerful dehydrating properties, low nucleophilicity of the phosphoric acid media, good solvation power, relatively mild acidity, and low oxidation potential. Even though PPA is a significantly weaker acid than H2SO4, it demonstrates 100% dehydration rate.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8017-16-1

Specifications

Melting Point -20°C
Boiling Point 530°C/1013 hPa
Density 2.06 g/cm3 at 20°C
Quantity 100 mL
Molecular Formula HO[P(OH)(O)O](n)H
MDL Number MFCD00084480
Synonym Polyphosphoric acid, PPA, Polyphosphoric acid
Grade Ungraded
Chemical Name or Material Polyphosphoric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.