Learn More
Vitlab™ Polyoxymethylene Forceps
Click to view available options
Length (Metric):
115 mm
145 mm
Unit Size:
Each
Pack of 5
Pack of 50
Description
- For bottling and decanting
- Yello
- Blunt
- Elastic
- Very good resilience
Specifications
Specifications
| Tip Style | Round |
| Material | POM |
| Length (Metric) | 115 mm |
| Product Type | Forceps |
| Quantity (No. per Case) | 5/Pack |
| For Use With (Application) | For autoclaving, bottling and decanting |
| Type | Forceps |