Learn More
Vitlab™ Polymethylpentene Forceps
Product Code. 10350881
Change view
Click to view available options
Length (Metric):
115 mm
145 mm
Unit Size:
Each
Pack of 10
Product Code. Length (Metric) unitSize
10350881 115 mm Each
11717202 115 mm Pack of 10
15751996 145 mm Pack of 10
3 options
Product Code. 10350881
Brand: Vitlab™ 67895
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured with PMP for very good resilience. VITLab™ Polymethylpentene Forceps are ideal for use in bottling and decanting.
  • White, Pointed, Elastic 
Specifications
Tip Style Pointed
Material PMP
Length (Metric) 115 mm
Product Type Forceps
Quantity (No. per Case) 10/Pack
For Use With (Application) For autoclaving and microwaves, bottling and decanting
Type Forceps