Learn More
Macherey-Nagel™ Standard SIL G Silica Layers on Polygram™ polyester sheets
Product Code. 10364501 Shop All Macherey-Nagel Products
Change view
Click to view available options
Dimensions (L x W):
2.5 x 7.5 cm
20 x 20 cm
4 x 8 cm
40 x 20 cm
5 x 20 cm
500 x 20 cm
Unit Size:
1 roll
Each
Pack of 200
Pack of 25
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
10364501 4 x 8 cm Pack of 50
10441193 20 x 20 cm Pack of 25
10515751 500 x 20 cm 1 roll
10640493 5 x 20 cm Pack of 50
12864491 2.5 x 7.5 cm Pack of 200
10516721 40 x 20 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10364501 Supplier Macherey-Nagel™ Supplier No. 805021
  • ONLINE EXCLUSIVE PRICE
    515.00 NOK
    396.88 NOK / Pack of 50
    Save 118.12 NOK
    22% Off
In Stock
Add to basket
Edge
Add to basket
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G silica layers on Polygram™ polyester sheets come in a variety of sizes. Select fluorescent indicator and choose from a variety of pack sizes.
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G silica layers on Polygram™ polyester sheets come in a variety of sizes. Select a fluorescent indicator and choose from a variety of pack sizes.
Polygram™ polyester sheets for TLC
  • Pre-coated ready to use plates
  • Broad range of plate sizes, surface coatings and backings
Absorbent
  • Silica 60
  • specific surface (BET) ∼ 500 m2/g
  • Mean pore size 60 Å
  • Specific pore volume 0.75 mL/g,
  • Particle size 5–17 μm
  • Standard grade
Thickness
  • For analytical plates 0.25 mm
  • For preparative plates 0.5 and 1 mm
  • For 2 mm preparative layers a slightly coarser material is used.
Inicators
  • Manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
  • Special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm)
Binders
  • Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents
Specifications
Pore Size 60 Å
Quantity 50 Pk.
Type Polyester Sheet
Fluorescent Indicator UV254
Pore Volume 0.75 mL/g
Thickness (Metric) 0.2 mm
Dimensions (L x W) 4 x 8 cm