Learn More
Macherey-Nagel™ Polygram CEL 300 PEI Polyester Sheets
Product Code. 10206861 Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
20 x 20 cm
Unit Size:
Pack of 25
Product Code. 10206861
Brand: Macherey-Nagel™ 801053
Only null left
Add to basket
Add to basket
TLC polyester sheets with fibrous cellulose impregnated with polyethyleneimine (PEI).
Specifications
Quantity 25 Pk.
Type Polyester Sheet
Thickness (Metric) 0.1 mm
Dimensions (L x W) 20 x 20 cm