Learn More
Macherey-Nagel™ Polygram Alox-25 UV254 Polyester Sheets
Brand:  Macherey-Nagel™ 807023
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10184661
  • 1915.00 NOK / Pack of 25
Estimated Shipment: 27-05-2024
Specifications
60 Å
Polyester Sheet
0.25 mm
25 Pk.
UV254
20 x 20 cm