Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- Column Packing
- Macherey-Nagel™ Polygosil™ Phase
Macherey-Nagel™ Polygosil™ Phase
Product Code. 10546301 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
10 g
100 g
Particle Size:
5 μm
7 μm
10 μm
Pore Size:
100 Å
1000 Å
300 Å
60 Å
Unit Size:
100g
10g
| Product Code. | Quantity | Particle Size | Pore Size | unitSize |
|---|---|---|---|---|
| 10546301 | 10 g | 7 μm | 100 Å | 10g |
| 10728232 | 100 g | 10 μm | 100 Å | 100g |
| 10699561 | 100 g | 10 μm | 60 Å | 100g |
| 10470882 | 100 g | 5 μm | 100 Å | 100g |
| 10337961 | 100 g | 5 μm | 60 Å | 100g |
| 10206651 | 100 g | 7 μm | 100 Å | 100g |
| 10431842 | 100 g | 7 μm | 60 Å | 100g |
| 10388481 | 100 g | 7 μm | 300 Å | 100g |
| 10398051 | 100 g | 7 μm | 1000 Å | 100g |
| 10012651 | 10 g | 10 μm | 100 Å | 10g |
| 10134741 | 10 g | 10 μm | 60 Å | 10g |
| 10072631 | 10 g | 5 μm | 100 Å | 10g |
| 10327961 | 10 g | 5 μm | 60 Å | 10g |
| 10062751 | 10 g | 7 μm | 1000 Å | 10g |
| 10378481 | 10 g | 7 μm | 300 Å | 10g |
| 10585771 | 10 g | 7 μm | 60 Å | 10g |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Particle Size | 7 μm |
| Pore Size | 100 Å |
| Technique | Liquid Chromatography |
| Quantity | 10 g |
| Type | Polygosil Phases |