Learn More
Polyethyleneimine on silica beads, anion exchange resin, benzylated, 40-200 mesh, Thermo Scientific™
Product Code. 11339458
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Unit Size:
10mL
250mL
50mL
Product Code. Quantity unitSize
11339458 10 mL 10mL
11319458 50 mL 50mL
11329458 250 mL 250mL
3 options
Product Code. 11339458
Brand: Thermo Scientific Alfa Aesar 043305.AB
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Solid phase organic synthesis, affinity chromatography, catalyst support
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9002-98-6
Molecular Formula ((C2H5N)zC2H4N)y(C2H5N)x
Molecular Weight (g/mol) 43.07
MDL Number MFCD00084427
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
Synonym ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
PubChem CID 9033
ChEBI CHEBI:30969
IUPAC Name aziridine
SMILES *-CCNCCN(-*)CCN-*
Specifications
Physical Form Powder
Quantity 10 mL
Mesh Size 40 to 200 Mesh
Sensitivity Hygroscopic
Chemical Name or Material Polyethyleneimine on silica beads anion exchange resin, Benzylated