Learn More
Kautex™ Polyethylene Wide Neck Jars
Product Code. 10430211
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
750 mL
1000 mL
1500 mL
2000 mL
Diameter (Metric) Inner Neck:
31 mm
41 mm
53 mm
67 mm
Unit Size:
Each
Pack of 108
Pack of 48
Pack of 700
Product Code. Capacity (Metric) Diameter (Metric) Inner Neck unitSize
10430211 100 mL 31 mm Each
17888117 100 mL 31 mm Pack of 700
10490781 250 mL 41 mm Each
10208850 500 mL 53 mm Each
10606741 750 mL 53 mm Each
17828117 750 mL 53 mm Pack of 108
10400981 1000 mL 67 mm Each
15991391 1500 mL 67 mm Pack of 48
10400791 1500 mL 67 mm Each
10370691 2000 mL 67 mm Each
10 options
Product Code. 10430211
Brand: Kautex™ 2000071492
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from HDPE for strength and durability. Kautex™ Polyethylene Wide Neck Jars provide easy to fill openings to minimize waste.
  • Round, sturdy, easy to clean and leakproof
  • Particularly suitable for storage, transport or disposal of powdery or granulated materials
  • Closures are not included
Applications:
  • packaging
  • dispensing
  • sending samples
  • mailing
  • transport
  • storage
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 100 mL
Color Natural
Diameter (Metric) Inner Neck 31 mm
For Use With (Application) Dispensing, Packaging, Storage
Material HDPE
No. per Case 720/Cs.
Weight (Metric) 0.01 kg
Closure Color null
Clarity Natural
Closure Included No
Mouth Wide
Shape Round
Height (Metric) 77 mm
Product Type Wide Neck Jar
Diameter (Metric) 48 mm