Learn More
Bel-Art™ Polyethylene Wash Bottle
Product Code. 17827906
Change view
Click to view available options
Mouth:
Narrow
Wide
Capacity (Metric):
125 mL
250 mL
Unit Size:
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mouth Capacity (Metric) unitSize
17827906 Narrow 125 mL Pack of 12
30193427 Wide 250 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17827906 Supplier Bel-Art™ Supplier No. F116180004
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
Low-density polyethylene wash bottles have color-coded polypropylene closures for easy identification and wide mouths for easy filling.
  • Enables quick, at-a-glance identification of bottle contents
  • Choose from Natural, Red, Yellow, Blue or Green closures
  • Bulk quantities available by special order
Specifications
Capacity (Metric) 125 mL
Capacity (English) 4 oz.
Color Natural
Diameter (Metric) Outer Neck 28 mm
Material Polyethylene, Polypropylene
Sterility Non-sterile
Closure Color Natural
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Narrow
Shape Cylindrical
Fit for Purpose Certified Quality
Product Type Wash Bottle
Chemical Label Blank
Closure Material Polypropylene