Learn More
GUERIN PLASTIQUES Polyethylene Sleeves
Product Code. 11816503
Click to view available options
Size:
0.1 Thick x 100Wmm
0.1 Thick x 120Wmm
0.1 Thick x 140Wmm
0.1 Thick x 160Wmm
0.1 Thick x 180Wmm
0.1 Thick x 200Wmm
0.1 Thick x 250Wmm
0.1 Thick x 300Wmm
0.1 Thick x 350Wmm
0.1 Thick x 55Wmm
0.1 Thick x 64Wmm
Unit Size:
Each
Product Code. 11816503
Brand: GUERIN PLASTIQUES 50149
Only null left
Add to basket
Add to basket
Use for food handling and general applications. GUERIN PLASTIQUES Polyethylene Sleeves are made of transparent, 0.1mm thick polyethylene.
  • 350m long in total
  • Available in a variety of widths
Specifications
Quantity 1/Pk.
Type Sleeve
Material Polyethylene
For Use With (Equipment) Food Quality
Length (Metric) 350 m
Size 0.1 Thick x 55Wmm
Color Transparent