Learn More
Kappes Systeme™ Polyethylene Storage Bin
Click to view available options
Color:
Gray
Unit Size:
Each
Description
Order Info
Color
Specifications
Specifications
| Type | Storage Bin |
| Material | Polyethylene |
| Color | Gray |
| Shape | Rectangular |
| For Use With | Feed and Roller Conveyor |
| For Use With (Application) | Food Storage |
| Length (Metric) | 500 mm |
| Width (Metric) | 300 mm |
| Height (Metric) | 250 mm |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.