Learn More
Masterflex™ Reducing Connector Barbed Fitting, HDPE
Product Code. 11820220
Change view
Click to view available options
For Use With:
1/2 x 1/4 in. tubing I.D.
1/4 x 3/16 in. tubing I.D.
3/8 x 1/4 in. tubing I.D.
5/8 x 1/2 in. tubing I.D.
5/8 x 3/8 in. tubing I.D.
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
11820220 1/2 x 1/4 in. tubing I.D. Pack of 10
11810220 3/8 x 1/4 in. tubing I.D. Pack of 10
11800220 1/4 x 3/16 in. tubing I.D. Pack of 10
11850220 5/8 x 1/2 in. tubing I.D. Pack of 10
11840220 5/8 x 3/8 in. tubing I.D. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11820220 Supplier Masterflex™ Supplier No. 3062238
Combine these simple, economical HDPE reducing connector with flexible tubing for use in your low-pressure applications (generally less than 30 psi). HDPE Reducing Connector Barbed Fittings slide into soft tubing. Tubing elasticity holds the tubing onto the fitting for low-pressure installations.
  • Designed for quick and easy assembly without tools
  • Very Good chemical resistance
  • Temp range: - 53 to 65°C
  • Sterilize by ethylene oxide
Specifications
Material Polyethylene
For Use With 1/2 x 1/4 in. tubing I.D.
Packaging Quantity 10 Pack
Product Type Barbed Fitting