Learn More
Masterflex™ Polyethylene Reducing Connector
Product Code. 11740119
Click to view available options
Tubing Size:
3/8 in. x 1/2 in.
Unit Size:
Pack of 100
Product Code. 11740119
Brand: Masterflex™ 3062642
Only null left
Add to basket
Add to basket
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools
  • Quick and easy assembly without tools
  • Just slip your flexible tubing or hose onto the barb to create a secure seal
  • Chemical resistant
  • Available in a variety of sizes
Specifications
Autoclavable Not Autoclavable
Material Polyethylene
Tubing Size 3/8 in. x 1/2 in.
Hose Barb 3/8 in. x 1/2 in.
Outlet Port 3/8 in.
Port Size 1/2 in.
Sterilization Method Ethylene Oxide
Color Translucent
Product Type Reducing Connector