Learn More
Hirschmann™ Polyethylene Poly Stopper
Description
- PE
- Available to fit a range of sizes
Specifications
Specifications
| Quantity | 1 Ea. |
| Standard Taper Joint | NS 14/23 |
| Material | Polyethylene |
| Product Type | Polyethylene Stopper |
| Shape | Hexagonal Grip |
| Sterility | Non-sterile |