Learn More
Brady™ Polyethylene PermaSleeve™ Wire Marker Sleeve
Specifications
Specifications
| Certifications/Compliance | UL Recognised to UL224; SAE-AMS-DTL-23053/5 C (Class 1); SAE-AS-81531 |
| Description | BMP71 PermaSleeve Wire Marker Sleeves |
| Color | Yellow |
| Finish | Matte |
| Material | Polyolefin |
| Printable Sides | Single-side Printable |
| Labels per Row | 1 |
| Quantity | 100/Pk. |
| Shape | Rectangle |
| Shrink Ratio | 3:01 |
| Show More |