Learn More
Pal International™ Deluxe Smooth Sole Fabric Overshoes
Product Code. 12943071
Click to view available options
Color:
White
Unit Size:
Pack of 200
Pack of 250
Product Code. 12943071
Brand: Pal International™ N68110
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for a better fit and improved comfort. Pal International™ Deluxe Smooth Sole Fabric Overshoes are especially suitable for labarotories or for visitors to hygienic manufacturing sites.
  • Manufactured from strong and durable non-woven polypropylene.
  • Higher spec overshoe designed for a better fit and improved comfort.
  • Especially suitable for more sensitive areas such as labarotories or for visitors to hygienic manufacturing sites.
  • Embossed sole gives extra grip and durability.
  • Elasticized band for easy wear and removal
  • DIsposable
  • One size fits all.
  • Package of 200
Specifications
Color White
Heel Style Elasticated ankle
Material Polypropylene
Size Universal
Sole Features Smooth
Product Type Overshoe