Learn More
Bel-Art™ Scienceware™ Narrow Mouth Polyethylene Wash Bottle
Product Code. p-8007856 Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
Unit Size:
Pack of 12
Product Code. 11704446
Brand: Bel-Art™ Scienceware™ F116180008
Only null left
Add to basket
Add to basket
Low-density polyethylene wash bottles offer different colors of polypropylene closures for quick, at-a-glance identification of contents
Specifications
Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Color Natural
Includes Natural Polypropylene Cap
Material Polyethylene
Quantity 12Pack
Sterility Non-sterile
Closure Color Natural
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Narrow
Shape Cylindrical
Product Type Wash Bottle
Chemical Label Unlabeled
Closure Material Polypropylene
Closure Size 28 mm
Closure Type Natural Polypropylene Cap