Learn More
Kartell™ Centrifuge Microtubes
Designed to withstand a centrifuge force up to RCF 11,000. Kartell™ Centrifuge Microtubes are ideal for sample analysis.
Brand:  Kartell™ 0129900
198.90 NOK valid until 2025-12-31
BEST PRICE on Promo! Use promo code "24328" to get your promotional price.
Product Code. 12049877
  • ONLINE EXCLUSIVE PRICE
    259.00 NOK
    258.16 NOK / Pack of 1000
    Save 0.84 NOK
    0.32% Off
Only null left
Estimated Shipment: 21-01-2026
Description
Designed to withstand a centrifuge force up to RCF 11,000. Kartell™ Centrifuge Microtubes are ideal for sample analysis.
  • Available with cap
  • Manufactured from polypropylene or polyethylene
  • Autoclavable
  • CE marked
  • IVD approved 