Learn More
Masterflex™ Polyethylene Male Pipe Adapter, pack of 100
Description
- Polyethylene Male Pipe Adapter
- Adapts from a NPT male thread to a hose barb
- Barbed fittings are designed for quick and easy assembly without tools
- Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
- Available in packs of ten
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | HDPE |
| Packaging Quantity | 100 Fittings |
| Tubing Size | 1/2 in. |
| Hose Barb | 1/2 in. |
| Outlet Port | 1/4 in. NPT(M) |
| Port Size | 1/2 in. |
| Sterilization Method | Ethylene Oxide |
| Thread Size | 1/4 in. NPT(M) |
| Shape | Straight |
| Show More |