Learn More
Masterflex™ Polyethylene Male Pipe Adapter, pack of 100
Description
- Polyethylene Male Pipe Adapter
- Adapts from a NPT male thread to a hose barb
- Barbed fittings are designed for quick and easy assembly without tools
- Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
- Available in packs of ten
Specifications
Specifications
| Description | Barb x NPT(M) Connection |
| Autoclavable | Not Autoclavable |
| Material | HDPE |
| Tubing Size | 3/8 in. |
| Hose Barb | 3/8 in. |
| Outlet Port | 3/8 in. NPT(M) |
| Port Size | 3/8 in. |
| Sterilization Method | Ethylene Oxide |
| Thread Size | 3/8 in. NPT(M) |
| Color | Translucent |
| Show More |