Learn More
Masterflex™ Polyethylene Male Pipe Adapter, pack of 100
Product Code. 11710119
Change view
Click to view available options
Tubing Size:
1/2 in.
3/8 in.
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size unitSize
11710119 3/8 in. Pack of 100
11780119 1/2 in. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11710119 Supplier Masterflex™ Supplier No. 3062562
Only null left
Add to basket
Add to basket
Designed for quick and easy assembly without tools
  • Polyethylene Male Pipe Adapter
  • Adapts from a NPT male thread to a hose barb
  • Barbed fittings are designed for quick and easy assembly without tools
  • Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
  • Available in packs of ten
Specifications
Description Barb x NPT(M) Connection
Autoclavable Not Autoclavable
Material HDPE
Tubing Size 3/8 in.
Hose Barb 3/8 in.
Outlet Port 3/8 in. NPT(M)
Port Size 3/8 in.
Sterilization Method Ethylene Oxide
Thread Size 3/8 in. NPT(M)
Color Translucent
Product Type Male Pipe Adapter