Learn More
Mettler Toledo™ Polyethylene LTS Pipette Tips
Click to view available options
Volume (Metric):
1000 μL
Unit Size:
Pack of 768
Specifications
Specifications
| Volume (Metric) | 1000 μL |
| Model | GPS-L1000 |
| For Use With (Application) | For Rainin LTS Pipettes |
| Format | Refill |
| For Use With (Equipment) | Rainin LTS |
| Material | Polyethylene |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.