Learn More
Buerkle™ Polyethylene Hose Couplings
Product Code. 10673443 Shop All Bürkle Products
Change view
Click to view available options
Diameter (Metric):
11 to 14 mm
13 to 16 mm
3 to 5 mm
5 to 7 mm
7 to 10 mm
9 to 12 mm
Unit Size:
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10673443 7 to 10 mm Pack of 10
10704433 3 to 5 mm Pack of 10
10742513 5 to 7 mm Pack of 10
10385512 9 to 12 mm Pack of 10
10161782 11 to 14 mm Pack of 10
10476892 13 to 16 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10673443 Supplier Buerkle™ Supplier No. 87040810
Only null left
Add to basket
Add to basket
Attachable, conical hose couplings for pressureless operation.
  • Manufactured from polyethylene
  • Provided 10 per pack
  • Available for a range of hose diameters
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EU Foodstuffs, RoHS
Diameter (Metric) 7 to 10 mm
Autoclavable Autoclavable
Material Polyethylene
For Use With 7 to 10 mm Hose
For Use With (Application) Pressureless Operation
Packaging Quantity 10/Pk.
Shape Conical
Product Type Hose Coupling