Learn More
Masterflex™ Polyethylene Elbow Pipe Adapter, pack of 100
Click to view available options
Diameter (English) Inner:
1/4 in.
Unit Size:
Pack of 100
Description
- Ideal for low-pressure applications
- Chemical resistant
- Available in a variety of sizes
- Economical barbed fittings easily connect fluid paths without the use of tools
- Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Specifications
| Description | Barb x NPT(M) Connection |
| Autoclavable | Not Autoclavable |
| Diameter (English) Inner | 1/4 in. |
| Material | HDPE |
| Overall Length (English) | 1 1/2 in. |
| Packaging Quantity | 100 Fittings |
| Tubing Size | 3/8 in. |
| Hose Barb | 3/8 in. |
| Outlet Port | 1/4 in. |
| Port Size | 3/8 in. |
| Show More |