Learn More
asecos™ Polyethylene Drum Chock
Product Code. 17816476 Shop All asecos Products
Change view
Click to view available options
Width (English):
2.95 in.
6.38 in.
Unit Size:
Each
Product Code. Width (English) unitSize
17816476 6.38 in. Each
17876586 2.95 in. Each
2 options
Product Code. 17816476
Brand: asecos™ 2802
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Polyethylene Drum Chock is ideal for the safe emptying of drums.
  • Seal made of EPDM
  • For caustic and aggressive chemicals
Specifications
Product Type Drum Faucet
Color Brown
For Use With (Equipment) 2 in. Drum
Material Polyethylene
Width (English) 6.38 in.
Width (Metric) 162.00 mm
Height (English) 6.14 in.
Height (Metric) 156.00 mm
Weight (English) 1.10 lb.
Weight (Metric) 0.5 kg