Learn More
Kartell™ Polyethylene Disc Membrane
Product Code. 10448462
Change view
Click to view available options
Diameter (Metric):
45 mm
55 mm
70 mm
80 mm
90 mm
110 mm
160 mm
240 mm
Unit Size:
Pack of 10
Product Code. Diameter (Metric) unitSize
10448462 70 mm Pack of 10
10214901 160 mm Pack of 10
12677765 110 mm Pack of 10
10001051 80 mm Pack of 10
12647785 240 mm Pack of 10
10011051 90 mm Pack of 10
10673852 45 mm Pack of 10
10224851 55 mm Pack of 10
8 options
Product Code. 10448462
Brand: Kartell™ 0083700
Only null left
Add to basket
Add to basket
These discs are made of polyethylene and have good chemical resistance. Kartell™ Polyethylene Disc Membrane is used to separate the filter paper from the funnel plate when working with a Buchner Funnel.
  • Useful interface between filter paper and perforated Buchner plate
  • Avoids clogging of filter holes
Specifications
Filter Type Screen Filter
Format Disc
Material Polyethylene
Pore Size 300 μm
For Use With (Application) Interface between filter paper and perforated Buchner plate
No. per Pack 10/Pack
Shape Circle
Diameter (Metric) 70 mm