Learn More
Buerkle™ Polyethylene Bung
Product Code. 15415574 Shop All Bürkle Products
Change view
Click to view available options
Standard Taper Joint:
10/19
12/21
14/23
19/26
24/29
29/32
34/35
45/40
7/16
Unit Size:
Each
Product Code. Standard Taper Joint unitSize
15415574 7/16 Each
11888172 19/26 Each
15445574 29/32 Each
11888182 34/35 Each
15376438 14/23 Each
11878182 45/40 Each
15337378 24/29 Each
15405574 10/19 Each
15435574 12/21 Each
9 options
Product Code. 15415574
Brand: Buerkle™ 97600716
Only null left
Add to basket
Add to basket
Manufactured with polyethylene for strength, durability and flexibility. Buerkle™ Polyethylene Bungs are available in a wide range of sizes.
  • Hollow stopper allows for use of tubes or funnels
  • Sealing rings provide a tight seal
  • Suitable for foodstuff
  • Octagonal shape facilitates easy opening
  • Polyethylene provides good chemical resistance
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For Foodstuffs
Standard Taper Joint 7/16
Color White, Red
Diameter (Metric) Bottom 5.9 mm
Diameter (Metric) Top 7.5 mm
Material Polyethylene
Product Type Bung