Learn More
Buerkle™ Polyethylene Bung
Product Code. 11888182 Shop All Bürkle Products
Click to view available options
Standard Taper Joint:
10/19
12/21
14/23
19/26
24/29
29/32
34/35
45/40
7/16
Unit Size:
Each
Product Code. 11888182
Brand: Buerkle™ 97603435
Only null left
Add to basket
Add to basket
Manufactured with polyethylene for strength, durability and flexibility. Buerkle™ Polyethylene Bungs are available in a wide range of sizes.
  • Hollow stopper allows for use of tubes or funnels
  • Sealing rings provide a tight seal
  • Suitable for foodstuff
  • Octagonal shape facilitates easy opening
  • Polyethylene provides good chemical resistance
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For Foodstuffs
Standard Taper Joint 34/35
Color White, Red
Diameter (Metric) Bottom 31 mm
Diameter (Metric) Top 34.5 mm
Material Polyethylene
Product Type Bung