Description
- Hollow stopper allows for use of tubes or funnels
- Sealing rings provide a tight seal
- Suitable for foodstuff
- Octagonal shape facilitates easy opening
- Polyethylene provides good chemical resistance
Specifications
Specifications
| For Use With (Application) | For Foodstuffs |
| Standard Taper Joint | 45/40 |
| Color | White, Red |
| Diameter (Metric) Bottom | 41 mm |
| Diameter (Metric) Top | 45 mm |
| Material | Polyethylene |
| Product Type | Bung |