Learn More
BRAND™ HDPE Bucket
Product Code. 10633972 Shop All Brand Products
Change view
Click to view available options
Unit Size:
Each
Capacity (Metric):
5 L
Height (Metric):
240 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size Capacity (Metric) Height (Metric)
10633972 Each - -
10613972 Each 5 L 240 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10633972 Supplier BRAND™ Supplier No. 71774
  • ONLINE EXCLUSIVE PRICE
    266.00 NOK
    200.22 NOK / Pack of 1
    Save 65.78 NOK
    24% Off
Estimated Shipment: 17-12-2026
Manufactured from High-density Polyethylene.BRAND™ HDPE Buckets include a handle, and graduation marks. Without spout. Push-on lid available for separate purchase.
  • Manufactured from High-density Polyethylene.
  • Graduated.
  • Includes handle.
  • Without spout.
  • Push-on lid available for separate purchase.
TRUSTED_SUSTAINABILITY
Specifications
Graduations Yes
Includes Handle