Learn More
Polyethylene Barrel Tap
Product Code. p-8010883
Encompass_Preferred
Change view
Click to view available options
Description:
2 in. (5.08 cm)
3/4 in. (1.905 cm)
Unit Size:
Each
Product Code. Description unitSize
13075623 2 in. (5.08 cm) Each
13065623 3/4 in. (1.905 cm) Each
2 options
Product Code. 13075623
Only null left
Add to basket
Add to basket
Two popular sized drum taps with efficient closing for safety purposes. They are particularly appropriate where containers are stored horizontally. Constructed in high density polyethylene they are resistant to a wide range of chemicals. The materials used meet FDA requirements. The 2 inch size is particularly useful for viscous liquids and for 205L drums, whereas the 3/4 inch tap fits smaller containers. In the case of the 2 inch tap, an adapter is offered which converts the BSP thread to Tri-sure for use with containers where this thread is fitted.
Specifications
Description 2 in. (5.08 cm)