Learn More
Masterflex™ Polyethylene Barbed Y-Connectors
Click to view available options
Tubing Size:
1/2 in.
1/8 in.
Unit Size:
Pack of 10
Description
- Barbed Y-Connectors
- Polyethylene
- Available in pack of ten
Specifications
Specifications
| Description | Barb Connections |
| Autoclavable | Not Autoclavable |
| Material | HDPE |
| Tubing Size | 1/2 in. |
| Hose Barb | 1/2 in. |
| Outlet Port | 1/2 in. |
| Port Size | 1/2 in. |
| Sterilization Method | Ethylene Oxide |
| Color | Translucent |
| Product Type | Barbed Y-Connector |