Learn More
Masterflex™ Polyethylene Barbed Y-Connectors
Product Code. 11770229
Click to view available options
Tubing Size:
1/2 in.
1/8 in.
Unit Size:
Pack of 10
Product Code. 11770229
Brand: Masterflex™ 3072603
Only null left
Add to basket
Add to basket
Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing
  • Barbed Y-Connectors
  • Polyethylene
  • Available in pack of ten
Specifications
Description Barb Connections
Autoclavable Not Autoclavable
Material HDPE
Tubing Size 1/8 in.
Hose Barb 1/8 in.
Outlet Port 1/8 in.
Port Size 1/8 in.
Sterilization Method Ethylene Oxide
Color Translucent
Product Type Barbed Y-Connector