Learn More
Bryson Packaging™ Polyethene Bags
Description
Polyethylene, resealable bags, clear or with three write-on bars for labelling. Ideal for storing stationery and sending samples etc.
Specifications
Specifications
| Closure Type | Grip Seal |
| Color | Clear |
| For Use With (Application) | Storing stationery |
| Material | Polyethylene |
| No. per Pack | 100 |
| Quantity | 100 Pack |
| Width (Metric) | 50 mm |
| Height (Metric) | 230 mm |
| Product Type | Bag |