Learn More
- Home
- Products
- Facility Safety and Maintenance
- Labels and Label Makers
- Custom Labels
- Brady™ Polyester Thermal-Transfer Printable Labels
Brady™ Polyester Thermal-Transfer Printable Labels
Product Code. 13009312
Change view
Click to view available options
Shape:
Rectangle
Rectangle with Rounded Corners
Round
Size (Metric):
101.6W x 50.8mmH
15.24 mm dia.
15.24 x 48.26 mm (W x H)
15.24 x 66.68 mm (W x H)
15.24W x 38.1 mmH
18.26W x 25.46 mmH
19 x 6.35 mm (W x H)
19W x 38.1 mmH
19W x 6.35 mmH
22.86W x 19 mmH
22.86W x 22.86 mmH
25.4 x 12.70 mm (W x H)
25.4W x 12.7mmH
25.4W x 4.75mmH
25.4W x 76.2mmH
25.4W x 9.53 mmH
31.75W x 19 mmH
34.93W x 45.72 mmH
38.10W x 6.35mmH
38.1W x 19 mmH
40W x 15 mmH
41.28 x 15.24 mm (W xH)
48.26W x 6.35 mmH
5.08W x 22.86 mmH
50.8 x 25.4 mm (W x H)
50.8W x 25.4 mmH
50.8W x 6.35 mmH
57.15W x 19 mmH
76.20W x 25.4mmH
76.20W x 50.8 mmH
76.2W x 25.4mmH
88.9W x 38.1 mmH
9.53 mm dia.
Finish:
Gloss
Matte
Satin
Adhesive Type:
Acrylic, Permanent
Rubber, Permanent
Unit Size:
1 pack
1 roll
| Product Code. | Shape | Size (Metric) | Finish | Adhesive Type | unitSize |
|---|---|---|---|---|---|
| 13009312 | Rectangle | 25.4W x 12.7mmH | Gloss | Rubber, Permanent | 1 pack |
| 13099262 | Rectangle | 101.6W x 50.8mmH | Gloss | Acrylic, Permanent | 1 roll |
| 10479883 | Round | 9.53 mm dia. | Matte | Acrylic, Permanent | 1 roll |
| 11504314 | Rectangle with Rounded Corners | 40W x 15 mmH | Matte | Acrylic, Permanent | 1 roll |
| 13019302 | Rectangle | 50.8W x 6.35 mmH | Gloss | Acrylic, Permanent | 1 roll |
| 11896023 | Rectangle | 19W x 38.1 mmH | Matte | Acrylic, Permanent | 1 roll |
| 13099302 | Rectangle | 38.10W x 6.35mmH | Gloss | Acrylic, Permanent | 1 roll |
| 11810855 | Rectangle | 19 x 6.35 mm (W x H) | Gloss | Acrylic, Permanent | 1 roll |
| 11816043 | Rectangle | 22.86W x 22.86 mmH | Matte | Acrylic, Permanent | 1 roll |
| 11583784 | Rectangle | 25.4W x 4.75mmH | Gloss | Acrylic, Permanent | 1 roll |
| 10209922 | Rectangle with Rounded Corners | 25.4W x 9.53 mmH | Matte | Acrylic, Permanent | 1 roll |
| 11836013 | Rectangle | 5.08W x 22.86 mmH | Matte | Acrylic, Permanent | 1 roll |
| 12834723 | Rectangle | 50.8W x 25.4 mmH | Satin | Acrylic, Permanent | 1 roll |
| 10020942 | Rectangle | 15.24 x 48.26 mm (W x H) | Matte | Acrylic, Permanent | 1 roll |
| 11538754 | Rectangle | 25.4W x 76.2mmH | Satin | Acrylic, Permanent | 1 roll |
| 11836023 | Rectangle | 34.93W x 45.72 mmH | Matte | Acrylic, Permanent | 1 roll |
| 11523724 | Rectangle | 76.20W x 25.4mmH | Gloss | Acrylic, Permanent | 1 roll |
| 10011932 | Rectangle with Rounded Corners | 15.24 mm dia. | Matte | Acrylic, Permanent | 1 roll |
| 11814714 | Rectangle | 76.20W x 50.8 mmH | Gloss | Acrylic, Permanent | 1 roll |
| 11503734 | Rectangle | 31.75W x 19 mmH | Gloss | Acrylic, Permanent | 1 roll |
| 11836043 | Rectangle | 38.1W x 19 mmH | Satin | Acrylic, Permanent | 1 roll |
| 10031932 | Rectangle | 15.24W x 38.1 mmH | Matte | Acrylic, Permanent | 1 roll |
| 10630134 | Rectangle | 50.8 x 25.4 mm (W x H) | Gloss | Acrylic, Permanent | 1 roll |
| 10315992 | Rectangle | 48.26W x 6.35 mmH | Matte | Acrylic, Permanent | 1 roll |
| 11885245 | Rectangle | 25.4 x 12.70 mm (W x H) | Gloss | Acrylic, Permanent | 1 roll |
| 10600784 | Rectangle | 41.28 x 15.24 mm (W xH) | Matte | Acrylic, Permanent | 1 roll |
| 11582715 | Rectangle | 76.2W x 25.4mmH | Gloss | Acrylic, Permanent | 1 roll |
| 10679343 | Rectangle | 88.9W x 38.1 mmH | Matte | Acrylic, Permanent | 1 roll |
| 10561515 | Rectangle | 18.26W x 25.46 mmH | Matte | Acrylic, Permanent | 1 roll |
| 11886013 | Rectangle | 15.24 x 66.68 mm (W x H) | Matte | Acrylic, Permanent | 1 roll |
| 12882699 | Rectangle | 22.86W x 19 mmH | Matte | Acrylic, Permanent | 1 roll |
| 11826033 | Rectangle | 57.15W x 19 mmH | Matte | Acrylic, Permanent | 1 roll |
| 11826043 | Rectangle | 19W x 6.35 mmH | Satin | Acrylic, Permanent | 1 roll |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Glossy white finish with permanent acrylic is thermal transfer printable
- RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
- Meets requirements of a halogen-free material per DIN VDE 0472 part 815
- Service temperature of -94 to 230° F (-70 to 110° C)
- Compatible printers: BBP11, BBP12, THT printer, 25 mm core
Specifications
Specifications
| Certifications/Compliance | RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC, UL Recognized, CSA Accepted |
| Description | Thermal Transfer Printable Labels |
| Adhesive Type | Rubber, Permanent |
| Coating | Powder |
| Color | White |
| Finish | Gloss |
| Format | Roll |
| Material | Polyester |
| Printable Sides | Single-side Printable |
| Print Technology | Thermal Transfer |
| Product Type | Thermal Transfer Printer Label |
| Sterility | Non-sterile |
| For Use With (Application) | Asset identification, rating and nameplate labels |
| For Use With (Equipment) | BBP81, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500 |
| Labels per Row | 1 |
| Min. Operating Temperature (Metric) | -40°C |
| Model | THT-59-483-10 |
| No. per Pack | 10000 Label(s) / Pack |
| Quantity | 10,000/Pk. |
| Shape | Rectangle |
| Size | 25.40 mm (W) x 12.70 mm (H) |
| Size (Metric) | 25.4W x 12.7mmH |
| Temperature Range (English) | -40°F to 248°F |
| Temperature Range (Metric) | -40°C to +120°C |
| Thickness (English) | 0.004 in. |
| Thickness (Metric) | 0.025 mm |
| Vertical Repeat (Metric) | 12.24 mm |
| Dimensions (LxW) | 12.70 x 25.40 mm |
| Length (English) | 0.5 in. |
| Width (English) | 1 in. |
| Width (Metric) | 25.4 mm |
| Width (English) Printable Area | 25.40 mm |
| Height (English) | 0.5 in. |
| Height (Metric) | 12.7 mm |
| Alternate Part Number | THT-59-483-10 |
| Show More Show Less |