Learn More
Brady™ Polyester Security Tags
Maintenance and quality Polyester Security Tags can be used for frequent inspection and of machinery and equipment.
272.00 NOK
Specifications
Product Type | Security Tag |
---|---|
Material | Polyester |
For Use With (Application) | Safety Identification |
Height (English) | 6.3 in. |
Height (Metric) | 160 mm |
Product Code | Brand | Displayed Text | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Displayed Text | Price | Quantity & Availability | |||||
11776845
|
Brady™
230820 |
Controle |
272.00 NOK
1 pack |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11746845
|
Brady™
230817 |
EN Reparat |
272.00 NOK
1 pack |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11786845
|
Brady™
230821 |
Controle Electrique |
272.00 NOK
1 pack |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11756845
|
Brady™
230818 |
Inspection |
272.00 NOK
1 pack |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11736845
|
Brady™
230816 |
Non-Accepted |
272.00 NOK
1 pack |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11766845
|
Brady™
230819 |
Quality Accepted |
272.00 NOK
1 pack |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11726845
|
Brady™
230815 |
Quality OK |
272.00 NOK
1 pack |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11706855
|
Brady™
230823 |
Refuse |
272.00 NOK
1 pack |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11796845
|
Brady™
230822 |
Reparation |
272.00 NOK
1 pack |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
- The reverse of the tag is blank.
Specifications
Security Tag | |
Safety Identification | |
160 mm | |
-40°F to +212°F | |
0.249 mm | |
75 mm |
Polyester | |
6.3 in. | |
10 Pk. | |
-40°C to +100°C | |
2.95 in. |