Learn More
Brady™ Polyester Printer Labels
Product Code. 11504274
Click to view available options
Size (Metric):
25.4 x 12.7 mm (W x L), 9.53 mm dia.
25.4 x 15.24 m (W x L)
48.26 mm x 15.24 m (W x L)
9.53 mm x 15.24 m (W x L)
Unit Size:
Each
Product Code. 11504274
Brand: Brady™ 710762
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
To identify frosted, frozen, or room temperature vials with overlap
  • Permanent acrylic adhesive
  • Opaque to cover legally protected patient information
  • Matt finish
Specifications
Description BMP71 FreezerBondz Label Printer Labels
Adhesive Type Permanent Acrylic
Color White
Finish Matte
Material Polyester
Product Type Printer Label
For Use With (Application) Laboratory identification
Labels per Row 1
Shape Rectangle with Rounded Corners
Size (Metric) 25.4 x 12.7 mm (W x L), 9.53 mm dia.
Dimensions (LxW) 12.70 x 25.40 mm
Diameter (Metric) 9.53 mm