Learn More
Brady™ Polyester Labels for Thermal Transfer Printers
Product Code. 30041400
Click to view available options
Size (Metric):
1.27 x 4.57 cm (W x H)
1.52 x 6.67 cm (W x H)
1.9 x 4.57 cm (W x H)
1.9 x 5.59 cm (W x H)
2.54 x 1.9 cm (W x H)
2.54 x 2.54 cm
6.35 W x 6.35 cm H
Finish:
Matte
For Use With (Equipment):
BBP11, BBP12, THT 25 mm Core Printers
BBP81, i5100, i6100, i7100, i7500, PR Plus, IP Series, THT 76 mm Core Printers
Quantity:
1500/Pk.
2500/Pk.
5000/Pk.
Unit Size:
1 roll
Pack of 2500
Product Code. 30041400
Brand: Brady™ 116410
Only null left
Add to basket
Add to basket
For barcoding and identification purposes
  • Available with gloss or matte finish
  • Available in different widths and roll lengths
Specifications
Certifications/Compliance RoHS 2002/95/EC
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Label
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Laboratory identification such as vials, centrifuge tubes, test tubes, straws, and slides
For Use With (Equipment) BBP11, BBP12, THT 25 mm Core Printers
Labels per Row 1
Min. Operating Temperature (Metric) -80°C
Model THT-142-481-2.5-SC
No. per Pack 2500 Label(s) / Roll
Quantity 2500/Pk.
Shape Rectangle
Size 63.50 mm (W) x 5.08 mm (H)
Size (Metric) 6.35 W x 6.35 cm H
Temperature Range (English) -112°F to +266°F
Temperature Range (Metric) -80°C to +130°C
Thickness (English) 0.0036 in.
Thickness (Metric) 0.0914 mm
Dimensions (LxW) 63.50 mm (W) x 5.08 mm (H)
Length (English) 2.5 in.
Width (English) 2.5 in.
Width (Metric) 6.35 cm
Width (English) Printable Area 63.50 mm
Height (English) 2.5 in.
Height (Metric) 6.35 cm
Alternate Part Number THT-142-481-2.5-SC