Learn More
Spex CertiPrep™ Polycarbonate vials
Product Code. 12839055 Shop All SPEX Certiprep Products
Click to view available options
Volume (Metric):
50 mL
Unit Size:
Pack of 48
Product Code. 12839055
Brand: Spex CertiPrep™ 2253PC48
This item requires a price update prior to being ordered.
Request a Price for this item here.
Polycarbonate cryovial with screw cap holds grinding media up to 1/2 inch and fits SPEX SamplePrep Homogeniser
Polycarbonate cryovial with screw-on cap, 290mm x 114mm, 50mL. Holds grinding media up to 1/2 in and fits SPEX SamplePrep homogeniser 2196-16-PE holder and 2664 cryo-block. Grinding load per vial 20mL. Supplied as a pack of 48 vials with 48 caps.
  • Holds grinding load of 20mL per vial
  • Polycarbonate so suitable for cryogenic grinding
Specifications
Material Polycarbonate
Volume (Metric) 50 mL
Includes with screw-on cap
Closure Type Screw Cap
Quantity 48 Pk.
For Use With (Equipment) Homogenizer