Learn More
Cole-Parmer™ Polycarbonate Tensioning Tools
Product Code. 11785049 Shop All Cole-Parmer Products
Change view
Click to view available options
Material:
Polycarbonate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
11785049 Polycarbonate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11785049 Supplier Cole-Parmer™ Supplier No. 0683007
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 18-12-2026
Adjust tension from light to very tight with this tool. Cole-Parmer™ Polycarbonate Tensioning Tool features dial-adjustable tension setting. Simply squeeze the handle to pull tie to a preset tension and cut.
Shock-resistant tool absorbs all impact. Ideal for 18 to 175 lb. cable or zip ties. No twisting necessary.
Specifications
Material Polycarbonate
For Use With (Application) Uniform clamping or work in series, equipped with a cutting device
No. per Pack 1 Ea.
Temperature Range (Metric) −40° to +85°C
Product Type Tensioning Tools