Learn More
Velp Scientifica™ Polycarbonate Tank
Product Code. 15349564 Shop All Velp Products
Change view
Click to view available options
Capacity (Metric):
5 L
7 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15349564 7 L Each
15339564 5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15349564 Supplier Velp Scientifica™ Supplier No. A00000091
Estimated Shipment: 28-12-2026
Velp Scientifica™ Polycarbonate Tank is a fundamental accessory of the Velp Scientifica™ (OCB) Open Circulating Bath.
  • Completely transparent for a constant visual control
Specifications
Capacity (Metric) 7 L
For Use With (Equipment) OCB
Type Tank
Material Polycarbonate