Learn More
3M™ 1610 Series Visitor Spectacles
Description
- Can be worn alone for general eye safety
- Lightweight polycarbonate lenses
- Molded side shields and brow guards
- Optical Class 1 for maximum clarity
- Acc. to ANSI Z87.1-2003
Specifications
Specifications
| Certifications/Compliance | EN166-1F |
| Product Type | Protective eyewear |
| For Use With (Application) | Protection against low energy impact, UV protection |
| Lens Material | Polycarbonate |
| UV Protection | Yes |