Learn More
Masterflex™ Polycarbonate Manifolds
Product Code. 11730559
Change view
Click to view available options
No. of Ports:
2
3
4
5
Includes:
180° Rotation
360° Rotation
Unit Size:
Each
Product Code. No. of Ports Includes unitSize
11730559 2 180° Rotation Each
11785239 4 180° Rotation Each
11700529 5 360° Rotation Each
11715279 3 360° Rotation Each
4 options
Product Code. 11730559
Brand: Masterflex™ 0646481
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulate flow without leaking with these manifolds with luer locks. Cole Parmer™ Polycarbonate Manifolds feature polyurethane stems, HDPE handles and female ports and luer locks.
  • Purchase manifolds individually or in convenient packs of ten
  • This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications
Specifications
Connection Type Luer Fitting
Description 180°
Includes 180° Rotation
Material Polycarbonate
No. of Ports 2