Learn More
Masterflex™ Polycarbonate Luer Fittings
Product Code. 11760279
Click to view available options
For Use With:
Female Luer x 1/4-28 UNF Thread
Female Luer x 1/8 in. Hose Barb
Female Luer x 10-32 in. UNF Thread
Male Luer w/ Lock Ring x 1/16 in. Hose Barb
Male Luer w/ Lock Ring x 1/4 in. Hose Barb
Male Luer w/ Lock Ring x 1/8 in. Hose Barb
Male Luer x 10-32 in. Thread
Male Luer x Female Luer Coupler
Male luer x male luer adapter
Unit Size:
Pack of 25
Product Code. 11760279
Brand: Masterflex™ 4550166
Designed to work with microtubing and luer-type syringes. Standard-sized connecting ends make these luer fittings completely interchangeable for surefire installations.
  • Manufactured from 100% virgin raw materials
  • Free of contaminants and ideal for syringes or small tubing
  • Choose from a range of sizes with a variety of connection options
  • Easily connect fluid paths without the use of tools
  • Male and female luers available
Specifications
Certifications/Compliance ISO 9001
Material Polycarbonate
For Use With Female Luer x 1/4-28 UNF Thread
For Use With (Application) Transitions between luer fittings and UNF or NPT threads
Packaging Quantity 25 Pack
Thread Size 1/4 to 28 UNF
Product Type Luer Fitting