Learn More
Masterflex™ Polycarbonate Luer Fittings
Click to view available options
For Use With:
Female Luer x 1/4-28 UNF Thread
Female Luer x 1/8 in. Hose Barb
Female Luer x 10-32 in. UNF Thread
Male Luer w/ Lock Ring x 1/16 in. Hose Barb
Male Luer w/ Lock Ring x 1/4 in. Hose Barb
Male Luer w/ Lock Ring x 1/8 in. Hose Barb
Male Luer x 10-32 in. Thread
Male Luer x Female Luer Coupler
Male luer x male luer adapter
Unit Size:
Pack of 25
Description
Designed to work with microtubing and luer-type syringes. Standard-sized connecting ends make these luer fittings completely interchangeable for surefire installations.
- Manufactured from 100% virgin raw materials
- Free of contaminants and ideal for syringes or small tubing
- Choose from a range of sizes with a variety of connection options
- Easily connect fluid paths without the use of tools
- Male and female luers available
Specifications
Specifications
| Certifications/Compliance | ISO 9001 |
| Material | Polycarbonate |
| For Use With | Female Luer x 1/8 in. Hose Barb |
| For Use With (Application) | For flexible tubing, reduce flow turbulence, semi-rigid tubing |
| Packaging Quantity | 25 Pack |
| Product Type | Luer Fitting |