Learn More
Corning™ Polycarbonate Erlenmeyer Flask with Dip Tube, Male Luer Lock, Sterile
Provide optimum visibility and durability
Brand:  Corning™ 11495
Additional Details : Weight : 0.86100kg
 View more versions of this product
Product Code. 17780688
  • 6039.01 NOK / Pack of 2
Estimated Shipment: 01-08-2024
Description
Corning™ closed system Erlenmeyer flasks will enhance the productivity of your shaker culture operations when adding or removing medium and cells. Flask is made from optically clear polycarbonate and is preassembled with an aseptic transfer cap. Unit is sterilized by gamma radiation and is nonpyrogenic and RNase-/DNase-free.
Flask:
  • Polycarbonate construction: USP Class VI material
  • Molded-in graduations for accuracy (+/-5%)
Aseptic Transfer Cap:
  • Chemically resistant, heat-sealable, heat-sealable, flexible tubing, 1/8” ID, 1/4” OD
  • Male Luer Lok Disconnect
Specifications
Specifications
101.4 oz.
Polypropylene
70 mm
Polycarbonate
Single-use
Sterile
Orange
With molded-in graduations
3 L
1/4 in. dip tube, male luer lock disconnect
Threaded
Aseptic Transfer Cap
Special Offers
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.