Learn More
Cytiva Polyamide Polypropylene Net Ring
Shop All Cytiva Products
Click to view available options
Volume (Metric):
20 μm
Unit Size:
Each
Specifications
Specifications
| Type | Net Ring |
| Volume (Metric) | 20 μm |
| Packaging Quantity | 5 Pack |
| For Use With | 16 mm I.D Columns |
| Material | Polyamide, Polypropylene |